4-Methoxy-N,N-dimethylaniline 98%

Supplier: Ambeed
Warning

A293646-10G A293646-1G A293646-250MG A293646-25G A293646-5G
77141-376EA 105.03 USD
77141-376 77141-378 77141-380 77141-382 77141-384
4-Methoxy-N,N-dimethylaniline 98%
4-Methoxy-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.206 g/mol
MDL Number: MFCD00155300
CAS Number: 701-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR