3-cyano-4-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-4-methoxybenzoic acid

A291767-1G A291767-250MG
77141-342EA 152.77 USD
77141-342 77141-344
3-cyano-4-methoxybenzoic acid 95%
3-cyano-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.157 g/mol
MDL Number: MFCD09271826
CAS Number: 117738-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR