6-Biotinamidohexanoic acid 95%

Supplier: Ambeed
Warning

A291733-250MG A291733-1G A291733-5G
77141-340EA 29.86 USD
77141-340 77571-798 77571-800
6-Biotinamidohexanoic acid 95%
6-Biotinamidohexanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₇N₃O₄S
MDL Number: MFCD00144853
CAS Number: 72040-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR