Ethyl 3-amino-1-(4-bromophenyl)-1H-pyrrole-2-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A290966-100MG A290966-250MG
77141-328EA 32.89 USD
77141-328 77141-330
Ethyl 3-amino-1-(4-bromophenyl)-1H-pyrrole-2-carboxylate hydrochloride 97%
Ethyl 3-amino-1-(4-bromophenyl)-1H-pyrrole-2-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrClN₂O₂
MW: 345.62 g/mol
MDL Number: MFCD26407208
CAS Number: 1272673-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR