Ethyl 7-bromoquinoline-2-carboxylate 97%

Supplier: Ambeed
Warning

A290423-100MG A290423-1G A290423-250MG
77141-318EA 77.45 USD
77141-318 77141-320 77141-322
Ethyl 7-bromoquinoline-2-carboxylate 97%
Ethyl 7-bromoquinoline-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂
MW: 280.11 g/mol
MDL Number: MFCD13190201
CAS Number: 1196153-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR