trans-2-Hexenal 98%

Supplier: Ambeed
Danger

Synonyms: (E)-2-Hexenal, trans-2-Hexenal

A289083-25G A289083-100G
77141-308EA 18.54 USD
77141-308 77616-248
trans-2-Hexenal 98%
trans-2-Hexenal

Precaución: for Research Use Only

Formula: C₆H₁₀O
MDL Number: MFCD00007008
CAS Number: 6728-26-3
UN: 1988
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR