(S)-Methyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 97%

Supplier: Ambeed
Warning

A286997-100MG A286997-1G A286997-250MG
77141-280EA 55.17 USD
77141-280 77141-282 77141-284
(S)-Methyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate 97%
(S)-Methyl 4-amino-2-((tert-butoxycarbonyl)amino)-4-oxobutanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₅
MW: 246.26 g/mol
MDL Number: MFCD08275816
CAS Number: 124842-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR