4'-Bromo-4-ethoxy-2,3-difluoro-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A286721-100MG A286721-1G A286721-250MG
77141-274EA 47.75 USD
77141-274 77141-276 77141-278
4'-Bromo-4-ethoxy-2,3-difluoro-1,1'-biphenyl 98%
4'-Bromo-4-ethoxy-2,3-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrF₂O
MW: 313.13 g/mol
MDL Number: MFCD25460355
CAS Number: 470461-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR