trans-5-Chloro-1-pentenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: (E)-(5-Chloropent-1-en-1-yl)boronic acid

A286585-100MG A286585-1G A286585-250MG
77141-268EA 30.77 USD
77141-268 77141-270 77141-272
trans-5-Chloro-1-pentenylboronic acid 98%
trans-5-Chloro-1-pentenylboronic acid

Precaución: for Research Use Only

Formula: C₅H₁₀BClO₂
MW: 148.3957 g/mol
MDL Number: MFCD01074658
CAS Number: 37490-32-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR