N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide dimethyl sulfoxide complex 98%

Supplier: Ambeed
Warning

Synonyms: Trametinib (DMSO solvate)

A286366-10MG A286366-5MG A286366-1G A286366-250MG A286366-1MG
77141-264EA 8.24 USD
77141-264 77141-266 77568-580 77616-226 77685-815
N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide dimethyl sulfoxide complex 98%
N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide dimethyl sulfoxide complex

Precaución: For research use only.

Formula: C₂₈H₂₉FIN₅O₅S
MDL Number: MFCD21609250
CAS Number: 1187431-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR