(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3R,7aS)-3-(Trichloromethyl)tetrahydropyrrolo[1,2-c]oxazol-1(3H)-one, (3R)-Trichloromethyl-cis-Tetrahydropyrrolo[1,2-c]oxazol-1-one, DK7774
A286130-10G
A286130-1G
A286130-5G
77141-258EA
39.14
USD
77141-258
77141-260
77141-262
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one 98%
(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
Precaución: for Research Use Only
Formula:
C₇H₈Cl₃NO₂ |
MDL Number:
MFCD08274464 CAS Number: 97538-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...