(R)-tert-Butyl (2-(4-bromophenyl)propyl)carbamate 97%

Supplier: Ambeed
Warning

A285183-100MG A285183-1G A285183-250MG A285183-5G
77141-228EA 7.21 USD
77141-228 77141-230 77141-232 77141-234
(R)-tert-Butyl (2-(4-bromophenyl)propyl)carbamate 97%
(R)-tert-Butyl (2-(4-bromophenyl)propyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrNO₂
MDL Number: MFCD28334223
CAS Number: 211315-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR