Methyl 2-amino-5-bromo-4-fluorobenzoate 96%

Supplier: Ambeed
Warning

A284077-1G A284077-250MG A284077-5G
77141-214EA 19.57 USD
77141-214 77141-216 77141-218
Methyl 2-amino-5-bromo-4-fluorobenzoate 96%
Methyl 2-amino-5-bromo-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
MDL Number: MFCD09951812
CAS Number: 1314987-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR