3-(Bromoacetyl)pyridine ydrobromide 97%

Supplier: Ambeed
Danger

Synonyms: 3-(2-Bromoacetyl)pyridine hydrobromide

A283219-1G A283219-25G A283219-5G
77141-208EA 10.61 USD
77141-208 77141-210 77141-212
3-(Bromoacetyl)pyridine ydrobromide 97%
3-(Bromoacetyl)pyridine ydrobromide

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 280.95 g/mol
MDL Number: MFCD00052182
CAS Number: 17694-68-7
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR