4-Chlorobenzo[d]thiazol-2(3H)-one 98%

Supplier: Ambeed
Warning

A282564-100MG A282564-1G A282564-250MG A282564-5G A282564-25G
77141-196EA 5.31 USD
77141-196 77141-198 77141-200 77141-202 77685-800
4-Chlorobenzo[d]thiazol-2(3H)-one 98%
4-Chlorobenzo[d]thiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄ClNOS
MW: 185.631 g/mol
MDL Number: MFCD00796306
CAS Number: 39205-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR