N,N'-((1R,2R)-1,2-Diphenylethane-1,2-diyl)bis(4-methylbenzenesulfonamide) ≥98%, ee 99%

Supplier: Ambeed
Warning

A282185-1G A282185-250MG A282185-5G
77141-188EA 40.32 USD
77141-188 77141-190 77141-192
N,N'-((1R,2R)-1,2-Diphenylethane-1,2-diyl)bis(4-methylbenzenesulfonamide) ≥98%, ee 99%
N,N'-((1R,2R)-1,2-Diphenylethane-1,2-diyl)bis(4-methylbenzenesulfonamide)

Precaución: for Research Use Only

Formula: C₂₈H₂₈N₂O₄S₂
MW: 520.66 g/mol
MDL Number: MFCD00269673
CAS Number: 121758-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR