tert-Butyl-N-(2-methoxyphenyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-methoxyphenyl)carbamate, 2-Methoxyaniline, N-BOC protected

A281595-25G A281595-1G A281595-5G
77141-186EA 389.36 USD
77141-186 77616-196 77616-198
tert-Butyl-N-(2-methoxyphenyl)carbamate 95%
tert-Butyl-N-(2-methoxyphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.27 g/mol
MDL Number: MFCD07127728
CAS Number: 154150-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR