5,7-Difluoroquinoline-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A280168-100MG A280168-250MG
77141-136EA 221.73 USD
77141-136 77141-138
5,7-Difluoroquinoline-6-carboxylic acid 97%
5,7-Difluoroquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₅F₂NO₂
MW: 209.14 g/mol
MDL Number: MFCD22573898
CAS Number: 1398504-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR