4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Carboxy-2-fluorophenylboronic acid pinacol ester
A279864-1G
A279864-250MG
A279864-5G
77141-130EA
104.02
USD
77141-130
77141-132
77141-134
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 97%
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₆BFO₄ |
MDL Number:
MFCD23098802 CAS Number: 2235384-35-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...