(R)-(-)-2-Aminooctane 98%

Supplier: Ambeed
Danger

Synonyms: (R)-(-)-1-Methylheptylamine, (R)-(-)-2-Octylamine, (R)-Octan-2-amine

A277625-1G A277625-250MG A277625-5G
77141-116EA 71.09 USD
77141-116 77141-118 77550-704
(R)-(-)-2-Aminooctane 98%
(R)-(-)-2-Aminooctane

Precaución: for Research Use Only

Formula: C₈H₁₉N
MW: 129.25 g/mol
MDL Number: MFCD01091017
CAS Number: 34566-05-7
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR