2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-trifluoromethylpyridin-3-bronic acid, pinacol ester

A277029-100MG A277029-1G A277029-250MG
77141-100EA 44.56 USD
77141-100 77141-102 77141-104
2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine 97%
2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₄BClF₃NO₂
MW: 307.5 g/mol
MDL Number: MFCD16996474
CAS Number: 2068737-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR