2-Cyano-4-methylpyridine 97%

Supplier: Ambeed
Danger

Synonyms: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

A276079-100G A276079-10G A276079-25G A276079-5G
77141-072EA 201.58 USD
77141-072 77141-074 77141-076 77141-078
2-Cyano-4-methylpyridine 97%
2-Cyano-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD00128868
CAS Number: 1620-76-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR