Boc-S-Tyr-OEt 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoate, Boc-Tyr-OEt

A275888-10G A275888-1G A275888-250MG
77141-052EA 21.22 USD
77141-052 77141-054 77141-056
Boc-S-Tyr-OEt 98%
Boc-S-Tyr-OEt

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MW: 309.36 g/mol
MDL Number: MFCD06795948
CAS Number: 72594-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR