1-(2-Chlorothiazol-5-yl)ethanone 98%

Supplier: Ambeed
Danger

A275756-100MG A275756-1G A275756-250MG
77141-046EA 12.74 USD
77141-046 77141-048 77141-050
1-(2-Chlorothiazol-5-yl)ethanone 98%
1-(2-Chlorothiazol-5-yl)ethanone

Precaución: for Research Use Only

Formula: C₅H₄ClNOS
MW: 161.61 g/mol
MDL Number: MFCD11847548
CAS Number: 885229-41-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR