(2,6-Difluoropyridin-3-yl)(phenyl)methanone 97%

Supplier: Ambeed
Danger

A273767-1G A273767-250MG
77141-022EA 239.77 USD
77141-022 77141-024
(2,6-Difluoropyridin-3-yl)(phenyl)methanone 97%
(2,6-Difluoropyridin-3-yl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₇F₂NO
MW: 219.18 g/mol
MDL Number: MFCD29922059
CAS Number: 1158955-21-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR