(+)-Taxifolin 98%

Supplier: Ambeed
Warning

A271339-100MG A271339-10MG A271339-1G A271339-1MG A271339-250MG A271339-50MG A271339-5G
77140-970EA 32.89 USD
77140-970 77140-972 77140-974 77140-976 77140-978 77140-980 77140-982
(+)-Taxifolin 98%
(+)-Taxifolin

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₇
MW: 304.2516 g/mol
MDL Number: MFCD00006845
CAS Number: 480-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR