(S)-2-(((tert-Butyldimethylsilyl)oxy)methyl)pyrrolidine 97%

Supplier: Ambeed
Warning

A270545-100MG A270545-1G A270545-250MG A270545-25G A270545-5G
77140-956EA 35.01 USD
77140-956 77140-958 77140-960 77140-962 77140-964
(S)-2-(((tert-Butyldimethylsilyl)oxy)methyl)pyrrolidine 97%
(S)-2-(((tert-Butyldimethylsilyl)oxy)methyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₂₅NOSi
MW: 215.40 g/mol
MDL Number: MFCD11502840
CAS Number: 134756-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR