(2-Fluoro-3-methoxy-5-(methoxycarbonyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A270363-1G A270363-250MG A270363-5G
77140-950EA 213.21 USD
77140-950 77140-952 77140-954
(2-Fluoro-3-methoxy-5-(methoxycarbonyl)phenyl)boronic acid 97%
(2-Fluoro-3-methoxy-5-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₅
MW: 227.98 g/mol
MDL Number: MFCD28101630
CAS Number: 2377610-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR