1400W dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1400W 2HCl, N-(3-(Aminomethyl)benzyl)acetimidamide dihydrochloride

A269504-10MG A269504-1MG A269504-50MG A269504-5MG
77140-932EA 80.34 USD
77140-932 77140-934 77140-936 77140-938
1400W dihydrochloride 95%
1400W dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃·2HCl
MW: 250.167 g/mol
MDL Number: MFCD03428622
CAS Number: 214358-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR