(5-Fluoro-2-methoxypyridin-3-yl)methanol 98%

Supplier: Ambeed
Warning

A267520-100MG A267520-250MG A267520-1G
77140-922EA 46.68 USD
77140-922 77140-924 77616-132
(5-Fluoro-2-methoxypyridin-3-yl)methanol 98%
(5-Fluoro-2-methoxypyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈FNO₂
MW: 157.142 g/mol
MDL Number: MFCD04972415
CAS Number: 874822-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR