(R)-1-Cbz-3-Boc-Aminopyrrolidine 97%

Supplier: Ambeed
Warning

A267117-1G A267117-5G
77140-912EA 86.53 USD
77140-912 77140-914
(R)-1-Cbz-3-Boc-Aminopyrrolidine 97%
(R)-1-Cbz-3-Boc-Aminopyrrolidine

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₄
MDL Number: MFCD08704349
CAS Number: 122536-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR