5-Chloro-2-nitrobenzyl alcohol 95%
Supplier: Ambeed
|
Warning
|
A266745-100G
A266745-10G
A266745-1G
A266745-250MG
A266745-25G
A266745-5G
77140-888EA
353.28
USD
77140-888
77140-890
77140-892
77140-894
77140-896
77140-898
5-Chloro-2-nitrobenzyl alcohol 95%
5-Chloro-2-nitrobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₃ MW: 187.58 g/mol |
MDL Number:
MFCD00007291 CAS Number: 73033-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...