(1S,3R)-(-)-Camphoric acid 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid

A266657-1G A266657-250MG A266657-5G
77140-882EA 59.42 USD
77140-882 77140-884 77140-886
(1S,3R)-(-)-Camphoric acid 98%
(1S,3R)-(-)-Camphoric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
MDL Number: MFCD00064937
CAS Number: 560-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR