3-Bromo-2-fluoro-4-methylpyridine 95%

Supplier: Ambeed
Warning

A266541-100MG A266541-250MG A266541-5G
77140-876EA 8.24 USD
77140-876 77140-878 77140-880
3-Bromo-2-fluoro-4-methylpyridine 95%
3-Bromo-2-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C6H5BrFN
MDL Number: MFCD08277289
CAS Number: 884495-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR