2-Nitro-4-(trifluoromethyl)benzyl alcohol 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Nitro-4-(trifluoromethyl)phenyl)methanol
A266023-1G
A266023-5G
77140-870EA
15.44
USD
77140-870
77140-872
2-Nitro-4-(trifluoromethyl)benzyl alcohol 95+%
2-Nitro-4-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO₃ |
MDL Number:
MFCD03095418 CAS Number: 133605-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...