Imatinib mesylate ≥99%

Supplier: Ambeed
Warning

A265691-100G A265691-100MG A265691-10G A265691-1G A265691-25G A265691-5G A265691-250MG A265691-1MG
77140-858EA 291.49 USD
77140-858 77140-860 77140-862 77140-864 77140-866 77140-868 77616-120 77685-758
Imatinib mesylate ≥99%
Imatinib mesylate

Precaución: for Research Use Only

Formula: C₂₉H₃₁N₇O·CH₄SO₃
MDL Number: MFCD04307699
CAS Number: 220127-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR