(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A265631-100MG A265631-1G A265631-250MG
77140-852EA 37.14 USD
77140-852 77140-854 77140-856
(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-4-yl)boronic acid 97%
(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₃
MW: 246.07 g/mol
MDL Number: MFCD22376771
CAS Number: 2056937-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR