3-Bromo-4-chlorotoluene 97%

Supplier: Ambeed
Warning

A265022-100G A265022-10G A265022-5G A265022-25G
77140-818EA 64.89 USD
77140-818 77140-820 77140-822 77685-755
3-Bromo-4-chlorotoluene 97%
3-Bromo-4-chlorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrCl
MDL Number: MFCD00079710
CAS Number: 57310-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR