Thieno[3,2-b]thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A264990-100MG A264990-1G A264990-250MG A264990-25G A264990-5G
77140-804EA 24.41 USD
77140-804 77140-806 77140-808 77140-810 77685-754
Thieno[3,2-b]thiophene-2-carboxylic acid 98%
Thieno[3,2-b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄O₂S₂
MDL Number: MFCD00115174
CAS Number: 1723-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR