5-Bromo-3,3-dimethyl-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A264724-100MG A264724-1G A264724-250MG A264724-5G
77140-796EA 82.76 USD
77140-796 77140-798 77140-800 77140-802
5-Bromo-3,3-dimethyl-2,3-dihydro-1H-inden-1-one 95%
5-Bromo-3,3-dimethyl-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO
MW: 239.11 g/mol
MDL Number: MFCD27927273
CAS Number: 67159-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR