4,6-Dibromo-o-toluidine 95%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dibromo-6-methylaniline, 2-Amino-3,5-dibromotoluene

A263870-10G A263870-1G A263870-5G
77140-790EA 32.89 USD
77140-790 77140-792 77140-794
4,6-Dibromo-o-toluidine 95%
4,6-Dibromo-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MW: 264.95 g/mol
MDL Number: MFCD00735681
CAS Number: 30273-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR