5-Bromosalicylamide 98%

Supplier: Ambeed
Warning

A263298-100G A263298-1G A263298-25G A263298-500G A263298-5G
77140-776EA 134.93 USD
77140-776 77140-778 77140-780 77140-782 77140-784
5-Bromosalicylamide 98%
5-Bromosalicylamide

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD00157694
CAS Number: 6329-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR