Ethyl 2-bromo-5-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A263149-1G A263149-25G A263149-5G
77140-770EA 22.66 USD
77140-770 77140-772 77140-774
Ethyl 2-bromo-5-(trifluoromethyl)benzoate 97%
Ethyl 2-bromo-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₃O₂
MDL Number: MFCD09999462
CAS Number: 1214336-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR