5-Chloro-2-methoxybenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

A263034-25G A263034-5G
77140-766EA 56.64 USD
77140-766 77140-768
5-Chloro-2-methoxybenzenesulphonyl chloride 98%
5-Chloro-2-methoxybenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O₃S
MDL Number: MFCD00052959
CAS Number: 22952-32-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR