(4-Bromo-3-methylisoxazol-5-yl)methyl acetate 97%

Supplier: Ambeed
Warning

A262561-1G A262561-250MG
77140-752EA 115.37 USD
77140-752 77140-754
(4-Bromo-3-methylisoxazol-5-yl)methyl acetate 97%
(4-Bromo-3-methylisoxazol-5-yl)methyl acetate

Precaución: for Research Use Only

Formula: C₇H₈BrNO₃
MDL Number: MFCD24387374
CAS Number: 1380089-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR