Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: DIchloro[bis(diphenylphosphinophenyl)ether]palladium(II)

A261801-10G A261801-1G A261801-250MG A261801-5G
77140-740EA 154.51 USD
77140-740 77140-742 77140-744 77140-746
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) 98%
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II)

Precaución: for Research Use Only

Formula: C₃₆H₂₈Cl₂OP₂Pd
MDL Number: MFCD09953446
CAS Number: 205319-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR