2-Fluoro-6-propoxypyridine-3-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-6-propoxypyridin-3-yl)boronic acid

A261545-1G A261545-250MG A261545-5G
77140-734EA 81.69 USD
77140-734 77140-736 77140-738
2-Fluoro-6-propoxypyridine-3-boronic acid 97%
2-Fluoro-6-propoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BFNO₃
MW: 198.987 g/mol
MDL Number: MFCD18434411
CAS Number: 2096338-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR