4-Chloro-2-methyl-6-nitroquinazoline 97%

Supplier: Ambeed
Warning

A260994-100MG A260994-250MG
77140-722EA 87 USD
77140-722 77140-724
4-Chloro-2-methyl-6-nitroquinazoline 97%
4-Chloro-2-methyl-6-nitroquinazoline

Precaución: for Research Use Only

Formula: C₉H₆ClN₃O₂
MW: 223.61 g/mol
MDL Number: MFCD11047030
CAS Number: 74151-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR