1,2-Benzisoxazol-3-amine 97%

Supplier: Ambeed
Warning

A260958-10G A260958-1G A260958-250MG A260958-5G
77140-710EA 101.97 USD
77140-710 77140-712 77140-714 77140-716
1,2-Benzisoxazol-3-amine 97%
1,2-Benzisoxazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MDL Number: MFCD03407353
CAS Number: 36216-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR