4-Hydroxy-3-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A258580-100MG A258580-10G A258580-1G A258580-250MG A258580-5G
77140-670EA 15.92 USD
77140-670 77140-672 77140-674 77140-676 77140-678
4-Hydroxy-3-iodobenzaldehyde 97%
4-Hydroxy-3-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅IO₂
MW: 248.0200 g/mol
MDL Number: MFCD00016983
CAS Number: 60032-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR